Learn More
Sigma Aldrich Fine Chemicals Biosciences Corning Ultra-Low attachment cell culture flasks, 24/cs
Catalog No. NC0979079
Encompass
Ultra-Low attachment 75cm2 rectangular canted cell culture flask, vent cap, sterile, 24/cs.
Catalog No. NC0979079 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS381424EA
Ultra-Low attachment 75cm2 rectangular canted cell culture flask, vent cap, sterile, 24/cs.