Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Universal CellSTACK™ Caps
BIOPHARMA

Catalog No. 0976227 Shop All Corning Products
Change view
Click to view available options
Closure Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
09-762-27 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-762-27 Supplier Corning™ Supplier No. 3969
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For filling, venting, and manipulation

  • Choice of 33mm polyethylene, solid or vented
  • Sterilized by gamma irradiation

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Black
Closure Material HDPE
Diameter (Metric) Outer Neck 3.3 cm
For Use With (Equipment) Used in closed systems or applications where venting is not necessary
Product Type Cap
Disposable Single-use
Sterility Sterile
Closure Type Screw
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.