Learn More
Corning™ Universal CellSTACK™ Caps
BIOPHARMA
Catalog No. 07201612 Shop All Corning Products
Change view
Click to view available options
Includes:
0.2μm PTFE membrane sealed into the peelable foil cover, vented overcap
Catalog No. Includes
07-201-612 0.2μm PTFE membrane sealed into the peelable foil cover, vented overcap
1 options
This item is not returnable. View return policy
Catalog No. 07-201-612 Supplier Corning™ Supplier No. 3732
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
For filling, venting, and manipulation
  • Designed to fit pre-existing filling systems for other product brands
  • 33 mm polyethylene threaded cap
  • Vented overcap has 0.2 μm PTFE membrane sealed into the peelable foil cover
  • Sterilized by gamma irradiation
  • Double bagged
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Orange
Closure Material HDPE
Diameter (Metric) Outer Neck 3.3 cm
For Use With (Equipment) Designed to fit pre-existing filling systems for other product brands
Includes 0.2μm PTFE membrane sealed into the peelable foil cover, vented overcap
Product Type Cap
Disposable Single-use
Sterility Sterile
Closure Type Screw, Vented
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.