Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Universal Fit Pipet Tips: Bulk Packed
GSA_VAGREENER_CHOICE

Catalog No. 07200296 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Tip Style:
Beveled, Round
Round
Packaging:
1000 Tips/Bag, 1 Bag/Cs.
1000 Tips/Bag, 10 Bags/Cs.
1000 Tips/Bag, 10,000 Tips/Cs.
1000 Tips/Bag, 1000 Tips/Cs.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Tip Style Packaging
07-200-296 1 to 200 μL Natural Beveled, Round 1000 Tips/Bag, 1000 Tips/Cs.
07-200-293 1 to 200 μL Natural Beveled, Round 1000 Tips/Bag, 10,000 Tips/Cs.
07-200-294 1 to 200 μL Yellow Round 1000 Tips/Bag, 10,000 Tips/Cs.
07-200-295 100 to 1000 μL Blue Round 1000 Tips/Bag, 10 Bags/Cs.
07-200-297 1 to 200 μL Yellow Round 1000 Tips/Bag, 1000 Tips/Cs.
07-200-298 100 to 1000 μL Blue Round 1000 Tips/Bag, 1 Bag/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07-200-296 Supplier Corning™ Supplier No. 4862
Only null left
Add to Cart
Add to Cart

Designed to assure a reliable fit with major brand pipetters

  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Natural
Format Bulk
Tip Style Beveled, Round
Packaging 1000 Tips/Bag, 1000 Tips/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.