Learn More
Corning™ UV-Transparent Microplates
BIOPHARMAGSA_VAGREENER_CHOICE
Catalog No. 07200623 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-200-623 Clear
1 options
Catalog No. 07-200-623 Supplier Corning™ Supplier No. 3635
Add to Cart
Add to Cart
Designed with a UV-transparent well bottom for determining protein and nucleic acid concentrations. Corning™ UV-Transparent Microplates allows UV absorbance readings with low background, especially at 260 to 280nm.
These microplates have a unique UV-transparent bottom, which is ideal for determining protein and/or nucleic acid concentrations. The UV-transparent bottom is molded to the top without adhesives for greater strength and maximum leak resistance. These microplates are for low background and consistent performance at 260 nm and 280 nm. Their broad linear detection range allows reliable detection of high and low concentrations of biomolecules.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified RNase/DNase-free
Lid Without Lid
Array 8 x 12
Packaging 2 inner packs of 25
Color Clear
Product Type UV Transparent Microplate
Material Plastic
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
For Use With (Application) Determining protein and/or nucleic acid concentrations, provides well-to-well uniformity, consistent low background and zero crosstalk
No. per Case 50/Cs.
No. per Pack 25/Pk.
Quantity 50/Cs.