Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon™ Vented Screw Caps for Flasks, Sterile
GREENER_CHOICE

Catalog No. p-176087 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Falcon™ 175 cm2 flasks
Falcon™ 25 cm2 flasks
Falcon™ 75 cm2 flasks
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
08-774-203 Falcon™ 25 cm2 flasks
08-774-204 Falcon™ 75 cm2 flasks
08-774-205 Falcon™ 175 cm2 flasks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-774-203 Supplier Falcon™ Supplier No. 354637
Only null left
Add to Cart
Add to Cart

Designed for use with Falcon 25-, 75-, and 175cm2 flasks

  • Ensure consistent gas exchange and minimize contamination
  • Polyethylene caps with an integral, hydrophobic 0.2 μm microporous membrane filter vent allow consistent gas exchange when the caps are in the closed position.
  • Gases required or cell growth and metabolism pass freely through the vent while microorganisms cannot. Falcon vented caps will not wet out.
  • Vented caps minimize contamination associated with standard open incubation. The vented caps prevent media that can become trapped in a partially opened cap from blocking gas exchange. The caps will not fall off in the incubator because of vibration.
TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polyethylene
Color Blue
For Use With (Equipment) Falcon™ 25 cm2 flasks
Includes 0.2μm Membrane vent
Product Type Cap, Vented
Sterility Sterile
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.