Learn More
Cole-Parmer CORNING XT COOLSINK 96U, EACH
Supplier:  Cole-Parmer 0439271
Encompass
Corning XT CoolSink 96U, each.Rapid temperature adaption to any temperature sourceReduce temperature variability and uncertainty with uniform well-to-well temperatureEnsured temperature accuracy of ±0.1° C across all tubes and plates for critical applicationsEasy to clean or sterilize
Catalog No. NC2026551