Learn More
Sigma Aldrich Fine Chemicals Biosciences Polypropylene Centrifuge Tubes | Conical Bottom with Flat Base | 50 mL and Other Sizes | Screw Cap or Plug Seal Closure | Sterile | Cell Culture Use | Each
Catalog No. NC2603873
Encompass
Polypropylene conical centrifuge tubes | Sterility: Sterile | Size: 50 mL and other sizes | Closure: Screw cap or plug seal | Tube shape: Conical bottom with flat base | Applications: Cell culture | Unit of Measure: Each
Catalog No. NC2603873 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS117062EA
Polypropylene conical centrifuge tubes | Sterility: Sterile | Size: 50 mL and other sizes | Closure: Screw cap or plug seal | Tube shape: Conical bottom with flat base | Applications: Cell culture | Unit of Measure: Each