Learn More
Cayman Chemical Verbenalin, Purity: ≥98%, CAS: 548-37-8, 10 mg
SDP
Catalog No. 502946293
Encompass_Preferred
An iridoid glucoside; inhibits the activities of COX-1 and COX-2 by 37 and 48%, respectively, at 258 µM; inhibits proliferation of AGS gastric, SF-268 CNS, HCT116 colon, NCI H460 lung, and MCF-7 breast cancer cells at 129 µM; reduces carrageen-induced paw edema and TPA-induced ear dema in mice; reduces increases in MDA levels and ROS production in isolated brain mitochondria isolated from, as well as decreases infarct volume and neurological deficits in, a rat model of MCAO-induced ischemia-reperfusion injury
Catalog No. 50-294-6293 Supplier Cayman Chemical Supplier No. 3466310 MG
An iridoid glucoside; inhibits the activities of COX-1 and COX-2 by 37 and 48%, respectively, at 258 µM; inhibits proliferation of AGS gastric, SF-268 CNS, HCT116 colon, NCI H460 lung, and MCF-7 breast cancer cells at 129 µM; reduces carrageen-induced paw edema and TPA-induced ear dema in mice; reduces increases in MDA levels and ROS production in isolated brain mitochondria isolated from, as well as decreases infarct volume and neurological deficits in, a rat model of MCAO-induced ischemia-reperfusion injury