Learn More
Waters Corp Waters Corona Discharge Pin, ASM (Sleeved), Ion Generation, MS System
Catalog No. 50466643 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement Corona Pin Assembly utilized in conjunction with an APCI probe as an alternative form of ion generation.
  • This is a replacement corona pin assembly.
  • It is utilized in conjunction with an APCI probe.
  • It serves as an alternative form of ion generation.
  • Typically associated with compatible mass spectrometry instruments.
Catalog No. 50-466-643 Supplier Waters Corp Supplier No. 700005346
This is a replacement Corona Pin Assembly utilized in conjunction with an APCI probe as an alternative form of ion generation.
  • This is a replacement corona pin assembly.
  • It is utilized in conjunction with an APCI probe.
  • It serves as an alternative form of ion generation.
  • Typically associated with compatible mass spectrometry instruments.