Learn More
Thermo Scientific Thermostat 2PCS
Catalog No. NC2042939 Shop All Thermo Scientific Products
Encompass
A sharp pointed metal wire needle electrode enclosed in a tube or cylinder produces a corona discharge electrical breakdown of the air gap for electrostatic applications like precipitation, ozone generation, painting, and separation when connected to a high voltage source.
Catalog No. NC2042939 Supplier Thermo Scientific Supplier No. 8011120683
A sharp pointed metal wire needle electrode enclosed in a tube or cylinder produces a corona discharge electrical breakdown of the air gap for electrostatic applications like precipitation, ozone generation, painting, and separation when connected to a high voltage source.