Learn More
Peak Scientific Instruments Ltd Corona Nitrogen 1010 Produces 5L/min of 98% pure Nitrogen
SDP
Catalog No. NC3364997
Encompass_Preferred
Suitable for applications requiring high purity nitrogenAvailable with or without an external compressorDedicated solution for Thermo Scientific's Corona and Vanquish instrumentsTried and tested membrane technology delivers laboratory-grade nitrogenGas is supplied on demand so generator works to your schedule 12 month comprehensive on-site warranty
Catalog No. NC3364997 Supplier Peak Scientific Instruments Ltd Supplier No. 814000
Add to Cart
Add to Cart
Request a Quote
Suitable for applications requiring high purity nitrogenAvailable with or without an external compressorDedicated solution for Thermo Scientific's Corona and Vanquish instrumentsTried and tested membrane technology delivers laboratory-grade nitrogenGas is supplied on demand so generator works to your schedule 12 month comprehensive on-site warranty