Learn More
Peak Scientific Instruments Ltd Corona Nitrogen 1010A 230V Produces 5L/min of 98% pure Nitrogen
SDP
Catalog No. 5000804193
Encompass_Preferred
Suitable for applications requiring high purity nitrogen
Full colour, touch-screen user interface for ease of operation
Built-in air compressor to maximize space in lab
Dedicated solution for ThermoFisher Scientific’s Corona and Vanquish instruments
Tried and tested membrane technology delivers high purity nitrogen
Gas is supplied on demand so generator works to your schedule
The most cost-effective method of gas supply
Catalog No. 50-008-04193 Supplier Peak Scientific Instruments Ltd Supplier No. 3304550
Suitable for applications requiring high purity nitrogen
Full colour, touch-screen user interface for ease of operation
Built-in air compressor to maximize space in lab
Dedicated solution for ThermoFisher Scientific’s Corona and Vanquish instruments
Tried and tested membrane technology delivers high purity nitrogen
Gas is supplied on demand so generator works to your schedule
The most cost-effective method of gas supply