Learn More
Waters Corp Corona Pin Assembly, ESCi, APCI, APPI Compatible
Catalog No. 50793368 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Corona Pin Assembly is a critical component for mass spectrometry instruments, enabling essential ionization techniques.
  • Required for ESCi, APCI, and dual-mode APCI/APPI operation.
  • Essential component for Xevo and Synapt mass spectrometry instruments.
Catalog No. 50-793-368 Supplier Waters Corp Supplier No. M956507BC1S
This Corona Pin Assembly is a critical component for mass spectrometry instruments, enabling essential ionization techniques.
  • Required for ESCi, APCI, and dual-mode APCI/APPI operation.
  • Essential component for Xevo and Synapt mass spectrometry instruments.