Learn More
Waters Corp WATERS CORPORATION Corona Pin Assembly, APCI Probe, MS System, Replacement
Catalog No. 50792877 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Corona Pin Assembly is a crucial replacement component utilized in mass spectrometry systems, specifically designed for ion generation in conjunction with an APCI probe. This assembly provides an alternative method for ion generation, ensuring continued optimal performance of your analytical instruments.
  • Functions as a replacement part
  • Used with APCI probe for alternative ion generation
  • Specific type is a corona pin
  • Designed for mass spectrometry system types
  • Comes with 1 unit per package
  • Requires replacement as needed
Catalog No. 50-792-877 Supplier Waters Corp Supplier No. 700003519
The Corona Pin Assembly is a crucial replacement component utilized in mass spectrometry systems, specifically designed for ion generation in conjunction with an APCI probe. This assembly provides an alternative method for ion generation, ensuring continued optimal performance of your analytical instruments.
  • Functions as a replacement part
  • Used with APCI probe for alternative ion generation
  • Specific type is a corona pin
  • Designed for mass spectrometry system types
  • Comes with 1 unit per package
  • Requires replacement as needed