Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coronarin D | 119188-37-3 | 5 MG
SDP

Catalog No. 5000420144
Encompass_Preferred

Coronarin D is a cytotoxic labdan-type diterpenes compound isolated from the rhizomes of H. coronarium.

  • Purity of 97.0%
  • Molecular weight of 318.45
  • Chemical formula C20H30O3
  • CAS number 119188-37-3
  • Appears as a white to off-white solid
  • Isolated from Hedychium coronarium Koen.
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • For research use only

Catalog No. 50-004-20144 Supplier Medchemexpress LLC Supplier No. HYN36305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coronarin D is a cytotoxic labdan-type diterpenes compound isolated from the rhizomes of H. coronarium.

  • Purity of 97.0%
  • Molecular weight of 318.45
  • Chemical formula C20H30O3
  • CAS number 119188-37-3
  • Appears as a white to off-white solid
  • Isolated from Hedychium coronarium Koen.
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.