Learn More
ZeptoMetrix Coronavirus (Strain: OC43) Culture Fluid (Heat Inactivated)
SDP
Catalog No. 22158319 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-319 1 mL
1 options
Catalog No. 22-158-319 Supplier ZeptoMetrix Supplier No. 0810024CFHI
Add to Cart
Add to Cart
Coronavirus (CoV) (Strain: OC43), a Group 2 CoV, is an enveloped virus with spiked projections, and contains a nucleocapsid with positive-sense RNA.
  • Heat inactivated viral culture fluids sold as consumable testing materials.
  • The suitability and performance characteristics should be determined by your laboratory for each intended usage.
  • These products are NOT intended for use in the manufacture or processing of injectable products subject to licensure under section 351 of the Public Health Service Act or for any other product intended for administration to humans.
  • Quality Control
Specifications
Content And Storage Heat inactivated viral culture fluids should be stored at -65™C or below
Description Repetitive freezing and thawing is not recommended (aliquot material if necessary).
Type Culture Fluid
Includes 1 mL of heat inactivated viral culture fluid
Quantity 1 mL
Strain Designations OC43