Learn More
Cell Signaling Technology Coronin 1A Monoclonal Antibody, Rabbit, Human and Mouse, 20 μl
SDP
Catalog No. 502730044
Encompass
The Coronin 1A (D6K5B) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total coronin 1A protein. Produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val200 of human coronin 1A, this antibody provides high specificity for its target and is not expected to cross-react with other members of the coronin protein family.
  • Validated for western blotting, immunohistochemistry (paraffin), immunofluorescence (frozen and immunocytochemistry), and flow cytometry (fixed/permeabilized) applications.
  • Reactivity demonstrated in human and mouse species.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended for research use only.
Catalog No. 50-273-0044 Supplier Cell Signaling Technology Supplier No. 92904T
The Coronin 1A (D6K5B) Rabbit Monoclonal Antibody is designed to recognize endogenous levels of total coronin 1A protein. Produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val200 of human coronin 1A, this antibody provides high specificity for its target and is not expected to cross-react with other members of the coronin protein family.
  • Validated for western blotting, immunohistochemistry (paraffin), immunofluorescence (frozen and immunocytochemistry), and flow cytometry (fixed/permeabilized) applications.
  • Reactivity demonstrated in human and mouse species.
  • Supplied in a formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Recommended for research use only.