Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Coronin 7 Peptide
SDP

Catalog No. 16596403
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-596-403 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-596-403 Supplier ProSci Supplier No. 6451P0050
Add to Cart
Add to Cart

Used for Blocking Activity

Coronin 7 peptide is used for blocking the activity of Coronin 7 antibody.

Specifications

Gene ID (Entrez) 79585
Species Human
Accession Number NP_078811
Common Name coronin7
Concentration 200 μg/mL
Content And Storage Store Coronin 7 peptide (at -20°C, stable for one year.)
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol CORO7
Quantity 0.05 mg
Sequence 19 amino acids near the carboxy terminus of human EI24.
Synonym CRN7, POD1, 0610011B16Rik, Coronin-7, 70 kDa WD repeat tumor rejection antigen homolog, Crn7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.