Learn More
Perkin Elmer US LLC 0.6 mm Corrosion-Resistant Probe
Catalog No. 509042740
Encompass_Preferred
Suitable for inorganic acids and most organic solvents,except NMP. Requires, but does not include, the capillary tubing assemblies.
Catalog No. 50-904-2740 Supplier Perkin Elmer US LLC Supplier No. B3000055
Suitable for inorganic acids and most organic solvents,except NMP. Requires, but does not include, the capillary tubing assemblies.