Learn More
TARGETMOL CHEMICALS INC CORRECTOR C4 5MG
SDP
Catalog No. 502580384
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Corrector C4 a corrector commonly used to study cystic fibrosis mutants works by alleviating the interaction between CFTR transmembrane domain mutants and protein homeostasis. purity: 98%
Catalog No. 50-258-0384 Supplier TARGETMOL CHEMICALS INC Supplier No. T310145MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Corrector C4 a corrector commonly used to study cystic fibrosis mutants works by alleviating the interaction between CFTR transmembrane domain mutants and protein homeostasis. purity: 98%