Learn More
Electron Microscopy Sciences Correlative Microscopy Coverslip 10x10 Grid, 1 mm
SDP
Supplier:  Electron Microscopy Sciences 6610803
Encompass_Preferred
A simple coverslip with a grid image that allows cells to grow and later be analyzed under brightfield or fluorescence microscopy before being further processed and sectioned for EM or other analysis.
Applications include Light Microscopy, Fluorescence, Scanning Electron Microscopy (SEM), Transmission Electron Microscopy (TEM), and High-Pressure Cryofixation.
Rectile film has been thoroughly tested with cell culture
Good growth of cell culture equivalent to conventional media
Good adhesion to the substrate without cell polylysine
Resistant to normal chemicals used in electron microscopy
Excellent transparency
Good optical quality in brightfield and UV fluorescence
Rigid and will not float in the middle of culture
Do not deform at high temperatures
Catalog No. 50-190-2447