Learn More
Bachem H-Val-His-OH
SDP
Supplier:  Bachem 4002910.0005
Encompass_Preferred
H-Val-His-OH, 13589-07-6, C11H18N4O3, Mr 254.29, 5g. Corresponds to the N-terminus of the b-chain of hemoglobin A which reacts with blood glucose yielding hemoglobin A1c.
Catalog No. 50-194-6844