Learn More
McMaster-Carr 430 Stainless Steel Compact Solenoid Valve with Exhaust Port
Catalog No. NC1470726
Encompass
430 Stainless Steel Compact Solenoid Valve with Exhaust Port Normally Closed, 24V DC, 1/4 NPT Female, 250 Maximum PSI
Catalog No. NC1470726 Supplier McMaster-Carr Supplier No. 7889K64
430 Stainless Steel Compact Solenoid Valve with Exhaust Port Normally Closed, 24V DC, 1/4 NPT Female, 250 Maximum PSI