Learn More
Wenesco CORROSION-RESISTANT HOT PLATE
Catalog No. NC1705405
Encompass
12x18 Anod Alum HP heated area 12x18plate dimensions 12x18x5
Catalog No. NC1705405 Supplier Wenesco Supplier No. HP1218CR
Add to Cart
Add to Cart
Request a Quote
12x18 Anod Alum HP heated area 12x18plate dimensions 12x18x5