Learn More
Perkin Elmer US LLC Probe Fitting Kit for S20 Series
Catalog No. 502016405
Encompass_Preferred
Corrosion-resistant probes are suitable for inorganic acids. Length: 10 in (25.4 cm) Probe Material: FEP-covered carbon fiber) Capillary Material: PFA) Includes: Probe with attached sample tubing) The contents of this kit are described as follows: 1.0 Mm Id Corrosion Resistant Carbon Fiber Probe (2 Blue Bands) (N0811956) 0.8 Mm Id Corrosion Resistant Carbon Fiber Probe (Red Band) (N0811953) 0.5 Mm Id Corrosion Resistant Carbon Fiber Probe (Blue Band) (N0777547)
Catalog No. 50-201-6405 Supplier Perkin Elmer US LLC Supplier No. N0830022
Corrosion-resistant probes are suitable for inorganic acids. Length: 10 in (25.4 cm) Probe Material: FEP-covered carbon fiber) Capillary Material: PFA) Includes: Probe with attached sample tubing) The contents of this kit are described as follows: 1.0 Mm Id Corrosion Resistant Carbon Fiber Probe (2 Blue Bands) (N0811956) 0.8 Mm Id Corrosion Resistant Carbon Fiber Probe (Red Band) (N0811953) 0.5 Mm Id Corrosion Resistant Carbon Fiber Probe (Blue Band) (N0777547)