Learn More
Thermo Scientific CORROSION-RESISTANT VACUUM PU
Supplier:  Thermo Scientific VLP200115
Encompass_Preferred
Pump; Oil Vacuum; VLP195FD series; Pump displacement 195L/min (at 60Hz); 162L/min (at 50Hz); Capacity 0.75L; 115V 60Hz
Catalog No. 50-870-637
Add to Cart
Request a Quote