Learn More
Thermo Scientific CORROSION-RESISTANT VACUUM PU
Catalog No. 50870641 Shop All Thermo Scientific Products
Encompass_Preferred
Pump; Oil Vacuum; Thermo Scientific; VLP120 series; Displacement: 283L/min (at 60Hz), 237L/min (at 50Hz); 6 x 17 x 9 in. (160 x 420 x 230mm); Ultimate Vacuum Level 1.5 mtorr/1.95 x10minus3 mbar; 115V 60Hz; 1L
Catalog No. 50-870-641 Supplier Thermo Scientific Supplier No. VLP285115
Add to Cart
Add to Cart
Request a Quote
Pump; Oil Vacuum; Thermo Scientific; VLP120 series; Displacement: 283L/min (at 60Hz), 237L/min (at 50Hz); 6 x 17 x 9 in. (160 x 420 x 230mm); Ultimate Vacuum Level 1.5 mtorr/1.95 x10minus3 mbar; 115V 60Hz; 1L