Learn More
Labelmaster Inc Corrosive Label, Worded, PVC-Free Film, 25/pk
SDP
Catalog No. NC0499948
Encompass_Preferred
Help take the guessing out of your shipping label solution with Labelmaster's Worded Corrosive Labels, revised to meet the design requirements of 49 CFR 172.407. In addition to complying with directives set by the DOT, these also meet the design and durability standards of 49 CFR, ICAO and other international air regulations.
Catalog No. NC0499948 Supplier Labelmaster Inc Supplier No. HMSL130S
Help take the guessing out of your shipping label solution with Labelmaster's Worded Corrosive Labels, revised to meet the design requirements of 49 CFR 172.407. In addition to complying with directives set by the DOT, these also meet the design and durability standards of 49 CFR, ICAO and other international air regulations.