Learn More
Grainger Eagle Acid Corrosive Safety Cabinet, 45 Gal, 2-Door, Self-Closing, Blue
Catalog No. 502763238
Encompass_Preferred
This 45 gal. (170.3 L) blue acid corrosive safety cabinet provides secure, compliant storage. The unit is constructed of 18-gauge steel with reinforced double-walled sides, top, bottom, and doors featuring a 1.5 in. (3.8 cm) insulating air space. The interior includes a 2 in. (5.1 cm) raised, leakproof door sill. Note: This cabinet is not recommended for use with sulfuric, nitric, or hydrochloric acids.
  • Self-closing, self-indexing doors with a 3-point stainless steel bullet latching system
  • Includes two 2 in. (5.1 cm) internal vents with fire baffles and caps
  • Two adjustable galvanized steel shelves, each with a 350 lb. (159 kg) uniform distributed load capacity
  • Poly shelf and sump trays included for added corrosion protection
  • Equipped with four adjustable galvanized leveling legs and grounding attachment
  • FM-approved and compliant with NFPA Code 30 and OSHA requirements
Catalog No. 50-276-3238 Supplier Grainger Supplier No. 15Y999
This 45 gal. (170.3 L) blue acid corrosive safety cabinet provides secure, compliant storage. The unit is constructed of 18-gauge steel with reinforced double-walled sides, top, bottom, and doors featuring a 1.5 in. (3.8 cm) insulating air space. The interior includes a 2 in. (5.1 cm) raised, leakproof door sill. Note: This cabinet is not recommended for use with sulfuric, nitric, or hydrochloric acids.
  • Self-closing, self-indexing doors with a 3-point stainless steel bullet latching system
  • Includes two 2 in. (5.1 cm) internal vents with fire baffles and caps
  • Two adjustable galvanized steel shelves, each with a 350 lb. (159 kg) uniform distributed load capacity
  • Poly shelf and sump trays included for added corrosion protection
  • Equipped with four adjustable galvanized leveling legs and grounding attachment
  • FM-approved and compliant with NFPA Code 30 and OSHA requirements