Learn More
Marketlab, Inc. Corrugated Polypropylene Freezer Box
Description
- Moisture-resistant to reduce the chance of mold growth
- Shipped flat packed for ease of storing
- Easy-to-assemble boxes come with preassembled dividers
- Store in temperatures as low as -80°C
- Able to freeze and thaw repeatedly
- For 1.5/2 mL tubes
Specifications
Specifications
| Type | Freezer Box |
| Color | Blue |