Learn More
Grainger Corrugated Tubing Nylon 1-1/4 in 50 ft
Catalog No. 502766228
Encompass_Preferred
The Corrugated Loom Tubing from Grainger is a flexible, corrugated tubing made of polyethylene that provides protection for wires, cables, and hoses. It has an inside diameter of 1 inch and is sold by the foot in lengths up to 100 feet. The tubing features a split seam for easy wrapping and securing with nylon cable ties. It is lightweight, moisture resistant, and rated for temperatures from -20°F to 176°F. The corrugated design allows the tubing to flex and bend without kinking. This loom tubing comes in black and is made in the USA. It can help organize and protect wires and cables in a wide variety of industrial, commercial, and residential applications.
Catalog No. 50-276-6228 Supplier Grainger Supplier No. 16Y266
The Corrugated Loom Tubing from Grainger is a flexible, corrugated tubing made of polyethylene that provides protection for wires, cables, and hoses. It has an inside diameter of 1 inch and is sold by the foot in lengths up to 100 feet. The tubing features a split seam for easy wrapping and securing with nylon cable ties. It is lightweight, moisture resistant, and rated for temperatures from -20°F to 176°F. The corrugated design allows the tubing to flex and bend without kinking. This loom tubing comes in black and is made in the USA. It can help organize and protect wires and cables in a wide variety of industrial, commercial, and residential applications.