Learn More
Grainger Corrugated Tubing Nylon 1/2 in 25 ft
Catalog No. 502765544
Encompass_Preferred
The Corrugated Loom Tubing 1/2" is a flexible, corrugated tubing made of polyethylene that provides protection for wires and cables. It has an inside diameter of 1/2", is sold by the foot, and is available in black, gray, and yellow colors. The tubing features a spiral cut design for easy wire insertion and has excellent abrasion resistance. It can withstand temperatures from -20°F to 176°F and is designed for both indoor and outdoor applications including wire harnesses, cable bundles, and anywhere that requires protecting wires from dust, moisture and abrasion. This tubing meets military specification MIL-T-21038 and is made in the USA.
Catalog No. 50-276-5544 Supplier Grainger Supplier No. 16Y264
The Corrugated Loom Tubing 1/2" is a flexible, corrugated tubing made of polyethylene that provides protection for wires and cables. It has an inside diameter of 1/2", is sold by the foot, and is available in black, gray, and yellow colors. The tubing features a spiral cut design for easy wire insertion and has excellent abrasion resistance. It can withstand temperatures from -20°F to 176°F and is designed for both indoor and outdoor applications including wire harnesses, cable bundles, and anywhere that requires protecting wires from dust, moisture and abrasion. This tubing meets military specification MIL-T-21038 and is made in the USA.