Learn More
Grainger Corrugated Tubing Nylon 1/2 in 100 ft
Catalog No. 502767003
Encompass_Preferred
The 1/2" Corrugated Loom Tubing by Electriduct is a flexible split loom tubing made of polyethylene and used to bundle and protect wires and cables. It has an inside diameter of 1/2", is sold by the foot, and is available in black color. The tubing features a corrugated exterior which allows it to easily flex and bend. It can withstand temperatures between -20°F to 176°F. The split design allows for quick and easy wire insertion without having to thread the ends. This abrasion resistant tubing helps prevent shorts and damaged wires. It can be used for a wide variety of wiring applications.
Catalog No. 50-276-7003 Supplier Grainger Supplier No. 16Y259
The 1/2" Corrugated Loom Tubing by Electriduct is a flexible split loom tubing made of polyethylene and used to bundle and protect wires and cables. It has an inside diameter of 1/2", is sold by the foot, and is available in black color. The tubing features a corrugated exterior which allows it to easily flex and bend. It can withstand temperatures between -20°F to 176°F. The split design allows for quick and easy wire insertion without having to thread the ends. This abrasion resistant tubing helps prevent shorts and damaged wires. It can be used for a wide variety of wiring applications.