Learn More
Grainger Corrugated Tubing Nylon 1/4 in 100 ft
Catalog No. 502767797
Encompass_Preferred
The 1/4" Corrugated Loom Tubing from Grainger is a flexible, split, corrugated tubing designed to protect wires, cables, and hoses. It is made from polyethylene and has an operating temperature range of -20°F to 176°F. The tubing has a 1/4" inside diameter and is sold by the foot in lengths up to 25 ft. It can be cut to desired length. The corrugated design allows the tubing to flex while still protecting contents. It is lightweight, moisture resistant, and resists abrasion. This tubing helps organize and protect wires and cables in various applications.
Catalog No. 50-276-7797 Supplier Grainger Supplier No. 16Y256
The 1/4" Corrugated Loom Tubing from Grainger is a flexible, split, corrugated tubing designed to protect wires, cables, and hoses. It is made from polyethylene and has an operating temperature range of -20°F to 176°F. The tubing has a 1/4" inside diameter and is sold by the foot in lengths up to 25 ft. It can be cut to desired length. The corrugated design allows the tubing to flex while still protecting contents. It is lightweight, moisture resistant, and resists abrasion. This tubing helps organize and protect wires and cables in various applications.