Learn More
Grainger Corrugated Tubing Nylon 1/4 in 25 ft
Catalog No. 502766857
Encompass_Preferred
The Grainger split corrugated loom tubing is a flexible plastic tubing used to bundle and protect wires and cables. It features a split down one side that allows for easy wire insertion without disconnecting. The tubing has a corrugated design which allows it to flex while still protecting contents. It is made from polyethylene and comes in sizes ranging from 1/4 inch to 2 inches in diameter. The tubing is UL recognized and rated for temperatures between -20°F to 176°F. It can be cut to desired lengths during installation. Overall, the split loom tubing is an easy to use option for bundling and protecting wires and cables.
Catalog No. 50-276-6857 Supplier Grainger Supplier No. 16Y261
The Grainger split corrugated loom tubing is a flexible plastic tubing used to bundle and protect wires and cables. It features a split down one side that allows for easy wire insertion without disconnecting. The tubing has a corrugated design which allows it to flex while still protecting contents. It is made from polyethylene and comes in sizes ranging from 1/4 inch to 2 inches in diameter. The tubing is UL recognized and rated for temperatures between -20°F to 176°F. It can be cut to desired lengths during installation. Overall, the split loom tubing is an easy to use option for bundling and protecting wires and cables.