Learn More
Grainger Corrugated Tubing Nylon 3/4 in 25 ft
Catalog No. 502765200
Encompass_Preferred
The 3/4" Corrugated Loom Tubing by Electriduct is a flexible plastic tubing designed to protect and organize wires and cables. It has an inner diameter of 0.75 inches and is made of polyethylene material. The tubing features a corrugated design which allows it to easily flex and bend. It is offered in a 25 foot length and black color. The operating temperature range is -20°F to 176°F. Potential applications include bundling computer cables, extension cords, wiring harnesses and more. The tubing helps prevent abrasion and tangling of wires while providing a neat, managed appearance.
Catalog No. 50-276-5200 Supplier Grainger Supplier No. 16Y265
The 3/4" Corrugated Loom Tubing by Electriduct is a flexible plastic tubing designed to protect and organize wires and cables. It has an inner diameter of 0.75 inches and is made of polyethylene material. The tubing features a corrugated design which allows it to easily flex and bend. It is offered in a 25 foot length and black color. The operating temperature range is -20°F to 176°F. Potential applications include bundling computer cables, extension cords, wiring harnesses and more. The tubing helps prevent abrasion and tangling of wires while providing a neat, managed appearance.