Learn More
Grainger Corrugated Tubing Nylon 3/4 in 100 ft
Catalog No. 502765060
Encompass_Preferred
The Corrugated Loom Tubing from Grainger is a flexible, corrugated tubing designed to protect wires, cables, and hoses. It is made from polyethylene and has an inside diameter of 1 inch. The tubing features a spiral cut design for easy installation and removal. It is rated for temperatures from -20°F to 176°F. The tubing is sold by the foot and is available in lengths up to 100 feet. It can provide abrasion resistance and protection from liquids and contaminants. The corrugated design allows the tubing to be bent without kinking or collapsing. It is a cost-effective option for protecting and organizing wires and cables.
Catalog No. 50-276-5060 Supplier Grainger Supplier No. 16Y260
The Corrugated Loom Tubing from Grainger is a flexible, corrugated tubing designed to protect wires, cables, and hoses. It is made from polyethylene and has an inside diameter of 1 inch. The tubing features a spiral cut design for easy installation and removal. It is rated for temperatures from -20°F to 176°F. The tubing is sold by the foot and is available in lengths up to 100 feet. It can provide abrasion resistance and protection from liquids and contaminants. The corrugated design allows the tubing to be bent without kinking or collapsing. It is a cost-effective option for protecting and organizing wires and cables.