Learn More
Grainger Corrugated Tubing Nylon 3/8 in 100 ft
Catalog No. 502767161
Encompass_Preferred
The Grainger Continuous Flex Corrugated Tubing is flexible, split, and designed to protect and organize wires and cables. Made from polyethylene, it withstands temperatures from -20°F to 176°F. With a smooth interior for easy wire pulling, it comes in sizes from 3/8" to 2" inside diameter and can be cut to length. Perforated sides allow easy splitting. UL recognized and CSA certified, it resists abrasion and protects from liquids and contaminants. Its corrugated design maintains shape while flexing, ideal for bundling wires in industrial, commercial, and residential settings.
Catalog No. 50-276-7161 Supplier Grainger Supplier No. 16Y258
The Grainger Continuous Flex Corrugated Tubing is flexible, split, and designed to protect and organize wires and cables. Made from polyethylene, it withstands temperatures from -20°F to 176°F. With a smooth interior for easy wire pulling, it comes in sizes from 3/8" to 2" inside diameter and can be cut to length. Perforated sides allow easy splitting. UL recognized and CSA certified, it resists abrasion and protects from liquids and contaminants. Its corrugated design maintains shape while flexing, ideal for bundling wires in industrial, commercial, and residential settings.