Learn More
Grainger Corrugated Tubing Nylon 3/8 in 25 ft
Catalog No. 502767004
Encompass_Preferred
The 3/4" Corrugated Loom Tubing from Grainger is a flexible, split, corrugated tubing designed to protect wires, cables, and hoses. It is made from polyethylene and has an operating temperature range of -20°F to 176°F. The tubing has a 3/4" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 1000'. It can be cut to desired length. The corrugated design allows the tubing to be bent to route wires and cables without kinking. It is designed for indoor and outdoor use and protects against abrasion, moisture, and chemicals. The split design allows for easy wire and cable insertion. It comes in black color.
Catalog No. 50-276-7004 Supplier Grainger Supplier No. 16Y263
The 3/4" Corrugated Loom Tubing from Grainger is a flexible, split, corrugated tubing designed to protect wires, cables, and hoses. It is made from polyethylene and has an operating temperature range of -20°F to 176°F. The tubing has a 3/4" inside diameter and is sold by the foot, with standard lengths ranging from 25' to 1000'. It can be cut to desired length. The corrugated design allows the tubing to be bent to route wires and cables without kinking. It is designed for indoor and outdoor use and protects against abrasion, moisture, and chemicals. The split design allows for easy wire and cable insertion. It comes in black color.