Learn More
Grainger Corrugated Tubing Nylon 5/16 in 100 ft
Catalog No. 502765197
Encompass_Preferred
The Corrugated Loom Tubing from Grainger is a flexible, split, corrugated loom tubing made of polyethylene. It has an inside diameter of 5/16 inches and is sold in 25 foot lengths. The tubing provides protection against abrasion and moisture for wires and cables. It has operating temperatures between -20°F to 176°F. The tubing meets military specification MIL-I-631D and is UL recognized to UL 224 and CSA C22.2 No. 198.1. It comes in black color.
Catalog No. 50-276-5197 Supplier Grainger Supplier No. 16Y257
The Corrugated Loom Tubing from Grainger is a flexible, split, corrugated loom tubing made of polyethylene. It has an inside diameter of 5/16 inches and is sold in 25 foot lengths. The tubing provides protection against abrasion and moisture for wires and cables. It has operating temperatures between -20°F to 176°F. The tubing meets military specification MIL-I-631D and is UL recognized to UL 224 and CSA C22.2 No. 198.1. It comes in black color.