Learn More
Grainger Corrugated Tubing PE 1-1/4 in 50 ft
Catalog No. 502767002
Encompass_Preferred
The Corrugated Loom Tubing 1-1/4" from Grainger is flexible polyethylene tubing that protects wires and cables. With a 1-1/4 inch inside diameter, it's sold by the foot in lengths from 25 to 100 feet. The flame retardant construction suits indoor and outdoor use. It can be cut with standard tools, and its corrugated design allows bending for routing cables. This tubing is an economical solution for bundling, protecting, and organizing wires in commercial and industrial settings.
Catalog No. 50-276-7002 Supplier Grainger Supplier No. 16Y255
The Corrugated Loom Tubing 1-1/4" from Grainger is flexible polyethylene tubing that protects wires and cables. With a 1-1/4 inch inside diameter, it's sold by the foot in lengths from 25 to 100 feet. The flame retardant construction suits indoor and outdoor use. It can be cut with standard tools, and its corrugated design allows bending for routing cables. This tubing is an economical solution for bundling, protecting, and organizing wires in commercial and industrial settings.