Learn More
Grainger Corrugated Tubing PE 1/2 in 100 ft
Catalog No. 502766186
Encompass_Preferred
The 1/2" Corrugated Loom Tubing by Electriduct is a flexible plastic tubing designed to protect and organize wires and cables. It features a corrugated design which allows it to easily flex and bend. The tubing has an inner diameter of 1/2" and is sold by the foot, with common lengths being 25, 50, and 100 foot rolls. It can withstand temperatures between -20°F to 176°F. This abrasion resistant vinyl tubing is UL recognized and CSA certified. It works well for running bundles of wire through tight spaces and providing protection from liquids, oils, and abrasion.
Catalog No. 50-276-6186 Supplier Grainger Supplier No. 16Y248
The 1/2" Corrugated Loom Tubing by Electriduct is a flexible plastic tubing designed to protect and organize wires and cables. It features a corrugated design which allows it to easily flex and bend. The tubing has an inner diameter of 1/2" and is sold by the foot, with common lengths being 25, 50, and 100 foot rolls. It can withstand temperatures between -20°F to 176°F. This abrasion resistant vinyl tubing is UL recognized and CSA certified. It works well for running bundles of wire through tight spaces and providing protection from liquids, oils, and abrasion.