Learn More
Grainger Corrugated Tubing PE 1/2 in 25 ft
Catalog No. 502766986
Encompass_Preferred
The Corrugated Loom Tubing 1/2" is a flexible, split, corrugated tubing made of polyethylene that can be used to bundle and protect wires and cables. It has an inside diameter of 1/2", is sold by the foot, and is available in black, gray or white colors. The tubing features a flame retardant split design for easy wire insertion and removal. It provides abrasion resistance and protection against liquids and chemicals. The operating temperature range is -20°F to 176°F. This tubing meets military specification MIL-I-631D and is made in the USA.
Catalog No. 50-276-6986 Supplier Grainger Supplier No. 16Y253
The Corrugated Loom Tubing 1/2" is a flexible, split, corrugated tubing made of polyethylene that can be used to bundle and protect wires and cables. It has an inside diameter of 1/2", is sold by the foot, and is available in black, gray or white colors. The tubing features a flame retardant split design for easy wire insertion and removal. It provides abrasion resistance and protection against liquids and chemicals. The operating temperature range is -20°F to 176°F. This tubing meets military specification MIL-I-631D and is made in the USA.