Learn More
Grainger Corrugated Tubing PE 1/4 in 100 ft
Catalog No. 502766977
Encompass_Preferred
The 3/4" Corrugated Loom Tubing from Grainger is a flexible, corrugated tubing designed to protect and organize wires and cables. It is made from polyethylene and has an inside diameter of 0.75 inches. The tubing features a spiral cut design for easy installation and removal. It is rated for temperatures from -20°F to 176°F. The tubing is sold by the foot, with common lengths ranging from 25 feet to 250 feet. It can be cut to desired lengths. The corrugated design allows the tubing to be bent without kinking while providing protection against abrasion, moisture and chemicals. It is commonly used for bundling and protecting wires and cables in various applications.
Catalog No. 50-276-6977 Supplier Grainger Supplier No. 16Y245
The 3/4" Corrugated Loom Tubing from Grainger is a flexible, corrugated tubing designed to protect and organize wires and cables. It is made from polyethylene and has an inside diameter of 0.75 inches. The tubing features a spiral cut design for easy installation and removal. It is rated for temperatures from -20°F to 176°F. The tubing is sold by the foot, with common lengths ranging from 25 feet to 250 feet. It can be cut to desired lengths. The corrugated design allows the tubing to be bent without kinking while providing protection against abrasion, moisture and chemicals. It is commonly used for bundling and protecting wires and cables in various applications.