Learn More
Grainger Corrugated Tubing PE 3/4 in 100 ft
Catalog No. 502766979
Encompass_Preferred
The 3/4" Corrugated Loom Tubing from Grainger is flexible, split, and corrugated to protect wires, cables, and hoses. Made from polyethylene, it operates between -20°F and 176°F. With a 3/4" inside diameter, it is sold by the foot in lengths from 25' to 1000' and can be cut as needed. The design allows bending for routing and is suitable for indoor and outdoor use, protecting against abrasion, moisture, and chemicals. The split design enables easy insertion of wires and cables without disconnecting fittings, ideal for various protection applications.
Catalog No. 50-276-6979 Supplier Grainger Supplier No. 16Y249
The 3/4" Corrugated Loom Tubing from Grainger is flexible, split, and corrugated to protect wires, cables, and hoses. Made from polyethylene, it operates between -20°F and 176°F. With a 3/4" inside diameter, it is sold by the foot in lengths from 25' to 1000' and can be cut as needed. The design allows bending for routing and is suitable for indoor and outdoor use, protecting against abrasion, moisture, and chemicals. The split design enables easy insertion of wires and cables without disconnecting fittings, ideal for various protection applications.