Learn More
Grainger Corrugated Tubing PE 3/4 in 25 ft
Catalog No. 502765540
Encompass_Preferred
The split corrugated loom tubing from Grainger is a flexible plastic tubing used to bundle and protect wires and cables. It features a split down one side that allows for easy wire insertion without disconnecting. The tubing has a corrugated design which allows it to flex while still protecting contents. It is made from polyethylene and comes in sizes ranging from 1/4 inch to 2 inches in diameter. The tubing is UL recognized and can withstand temperatures from -20°F to 176°F. It comes in standard lengths of 25, 50, and 100 feet.
Catalog No. 50-276-5540 Supplier Grainger Supplier No. 16Y254
The split corrugated loom tubing from Grainger is a flexible plastic tubing used to bundle and protect wires and cables. It features a split down one side that allows for easy wire insertion without disconnecting. The tubing has a corrugated design which allows it to flex while still protecting contents. It is made from polyethylene and comes in sizes ranging from 1/4 inch to 2 inches in diameter. The tubing is UL recognized and can withstand temperatures from -20°F to 176°F. It comes in standard lengths of 25, 50, and 100 feet.