Learn More
Grainger Corrugated Tubing PE 3/8 in 100 ft
Catalog No. 502765056
Encompass_Preferred
The Grainger 3/8" Corrugated Polyethylene Tubing is a flexible tubing made from polyethylene plastic with a corrugated exterior. It has an inside diameter of 3/8 inches and comes in 100 foot rolls. The tubing is lightweight, flexible, crush resistant, and chemical resistant. It has operating temperature range of -50°F to 160°F. The tubing is commonly used for protecting wires and cables from abrasion. This particular tubing product has a yellow exterior and is sold by Grainger, product number GGE16Y247.
Catalog No. 50-276-5056 Supplier Grainger Supplier No. 16Y247
The Grainger 3/8" Corrugated Polyethylene Tubing is a flexible tubing made from polyethylene plastic with a corrugated exterior. It has an inside diameter of 3/8 inches and comes in 100 foot rolls. The tubing is lightweight, flexible, crush resistant, and chemical resistant. It has operating temperature range of -50°F to 160°F. The tubing is commonly used for protecting wires and cables from abrasion. This particular tubing product has a yellow exterior and is sold by Grainger, product number GGE16Y247.