Learn More
Grainger Corrugated Tubing PE 3/8 in 25 ft
Catalog No. 502765539
Encompass_Preferred
The Grainger 16Y252 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron body with standard mounting feet and comes pre-wired with color coded leads. This general purpose motor can be used for a wide variety of industrial, commercial and residential applications.
Catalog No. 50-276-5539 Supplier Grainger Supplier No. 16Y252
The Grainger 16Y252 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron body with standard mounting feet and comes pre-wired with color coded leads. This general purpose motor can be used for a wide variety of industrial, commercial and residential applications.