Learn More
Grainger Corrugated Tubing PE 5/16 in 100 ft
Catalog No. 502766210
Encompass_Preferred
Grainger offers corrugated loom tubing for wire and cable management. The tubing is made of flexible PVC and comes in sizes ranging from 1/4 inch to 2 inches in diameter. It is designed to protect and organize wires and cables. The tubing has a corrugated design which allows it to easily flex and bend. It is UL recognized and rated for temperatures from -20°F to 176°F. The tubing comes in standard lengths of 25, 50, and 100 feet and in various colors including black, white, gray, yellow, red, and blue. Grainger corrugated loom tubing provides an affordable and effective solution for bundling, routing, and protecting wires and cables.
Catalog No. 50-276-6210 Supplier Grainger Supplier No. 16Y246
Grainger offers corrugated loom tubing for wire and cable management. The tubing is made of flexible PVC and comes in sizes ranging from 1/4 inch to 2 inches in diameter. It is designed to protect and organize wires and cables. The tubing has a corrugated design which allows it to easily flex and bend. It is UL recognized and rated for temperatures from -20°F to 176°F. The tubing comes in standard lengths of 25, 50, and 100 feet and in various colors including black, white, gray, yellow, red, and blue. Grainger corrugated loom tubing provides an affordable and effective solution for bundling, routing, and protecting wires and cables.